Search
Search

Mlc1| Gene Symbol: Mlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300640.1 | NP_001287569.1 | 2 - 3 | 98 | 60 | |
| NM_057291.4 | NP_476639.1 | 2 - 3 | 98 | 60 | ||
| NM_057292.4 | NP_476640.1 | 2 - 3 | 98 | 60 | ||
| GenBank mRNA | AY070972.1 | - | 2 - 3 | 115 | 60 | |
| K01567.1 | - | 2 - 3 | 76 | 60 | ||
Set Membership: |
|