Search
Search

slou| Gene Symbol: slou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275879.1 | NP_001262808.1 | 1 - 2 | 1550 | 70 | |
| NM_057309.3 | NP_476657.1 | 1 - 2 | 1550 | 70 | ||
| GenBank mRNA | X55393.1 | - | 1551 | 70 | ||
Set Membership: |
|