Search
Search

tws| Gene Symbol: tws | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300340.1 | NP_001287269.1 | 3 - 4 | 1474 | 81 | |
| NM_057532.4 | NP_476880.1 | 4 - 5 | 1653 | 81 | ||
| NM_057533.3 | NP_476881.1 | 4 - 5 | 1437 | 81 | ||
| NM_134284.3 | NP_599111.1 | 4 - 5 | 1380 | 81 | ||
| NM_134285.2 | NP_599112.1 | 4 - 5 | 1375 | 81 | ||
| NM_134286.3 | NP_599113.1 | 4 - 5 | 1512 | 81 | ||
| NM_169329.2 | NP_731451.1 | 4 - 5 | 1282 | 81 | ||
| NM_169330.2 | NP_731452.1 | 4 - 5 | 1540 | 81 | ||
| NM_169331.2 | NP_731453.1 | 4 - 5 | 1433 | 81 | ||
| GenBank mRNA | AY061152.1 | - | 4 - 5 | 1629 | 81 | |
| BT099776.1 | - | 4 - 5 | 1433 | 81 | ||
| L07581.1 | - | 4 - 5 | 1487 | 81 | ||
Set Membership: |
|