Search
Search

Doa| Gene Symbol: Doa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014678.3 | NP_001014678.1 | 3 - 4 | 2167 | 75 | |
| NM_001014680.4 | NP_001014680.1 | 4 - 5 | 883 | 75 | ||
| NM_001014682.3 | NP_001014682.1 | 4 - 5 | 759 | 75 | ||
| NM_001043300.3 | NP_001036765.1 | 4 - 5 | 883 | 75 | ||
| NM_001144648.3 | NP_001138120.1 | 5 - 6 | 826 | 75 | ||
| NM_001144649.2 | NP_001138121.1 | 5 - 6 | 826 | 75 | ||
| NM_001144650.1 | NP_001138122.1 | 4 - 5 | 759 | 75 | ||
| GenBank mRNA | AY061474.1 | - | 4 - 5 | 877 | 75 | |
| BT133441.1 | - | 3 - 4 | 2165 | 75 | ||
| EF489455.1 | - | 3 - 4 | 2165 | 75 | ||
Set Membership: |
|