Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02134961_g1
          See other DOA GE Assays ›
          Gene Symbol:
          Doa
          Gene Name:
          Darkener of apricot
          Gene Aliases:
          0844/02, CG1658, CG31049, CG33204, CG33553, CG42320, DOA, DOA/CLK2, Dmel\CG42320, Dmel_CG42320, Msu, doa, l(3)01705, l(3)S084402, l(3)s2784
          Chromosome Location:
          Chr.3R: 28888073 - 28922772 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0265998
          Assay ID Dm02134961_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Doa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014678.3 NP_001014678.1 5 - 6 2302 79
          NM_001014679.2 NP_001014679.1 3 - 4 1665 79
          NM_001014680.4 NP_001014680.1 6 - 7 1018 79
          NM_001014681.4 NP_001014681.2 5 - 6 5283 79
          NM_001014682.3 NP_001014682.1 6 - 7 894 79
          NM_001043298.2 NP_001036763.2 4 - 5 3728 79
          NM_001043299.3 NP_001036764.2 7 - 8 1101 79
          NM_001043300.3 NP_001036765.1 6 - 7 1018 79
          NM_001144648.3 NP_001138120.1 7 - 8 961 79
          NM_001144649.2 NP_001138121.1 7 - 8 961 79
          NM_001144650.1 NP_001138122.1 6 - 7 894 79
          NM_001260424.1 NP_001247353.1 4 - 5 1939 79
          NM_001260425.1 NP_001247354.1 5 - 6 1930 79
          NM_001382085.1 NP_001368951.1 7 - 8 1158 79
          NM_176581.4 NP_788758.3 5 - 6 2692 79
          GenBank mRNA AY061474.1 - 6 - 7 1012 79
          AY069573.1 - 3 - 4 1665 79
          BT133441.1 - 5 - 6 2300 79
          EF489455.1 - 5 - 6 2300 79

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          regulation of alternative mRNA splicing, via spliceosome
          positive regulation of antimicrobial peptide production
          protein phosphorylation
          blastoderm segmentation
          nervous system development
          sex differentiation
          protein secretion
          positive regulation of autophagy
          somatic sex determination
          compound eye photoreceptor development
          positive regulation of Toll signaling pathway
          protein autophosphorylation
          compound eye development
          negative regulation of male germ cell proliferation
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline