Search
Search

cno| Gene Symbol: cno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275342.1 | NP_001262271.1 | 14 - 15 | 5312 | 76 | |
| NM_001275343.1 | NP_001262272.1 | 17 - 18 | 6292 | 76 | ||
| NM_001275344.1 | NP_001262273.1 | 18 - 19 | 6343 | 76 | ||
| NM_001275345.1 | NP_001262274.1 | 20 - 21 | 8723 | 76 | ||
| NM_001316565.1 | NP_001303494.1 | 19 - 20 | 6592 | 76 | ||
| NM_079508.4 | NP_524232.2 | 19 - 20 | 6592 | 76 | ||
| NM_169025.3 | NP_730891.1 | 16 - 17 | 6080 | 76 | ||
| GenBank mRNA | BT044227.1 | - | 19 - 20 | 6561 | 76 | |
Set Membership: |
|