Search
Search

Vha26| Gene Symbol: Vha26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300253.1 | NP_001287182.1 | 1 - 2 | 276 | 67 | |
| NM_079513.3 | NP_524237.1 | 2 - 3 | 178 | 67 | ||
| NM_169073.2 | NP_730957.1 | 1 - 2 | 276 | 67 | ||
| GenBank mRNA | AF145618.1 | - | 1 - 2 | 221 | 67 | |
| U38198.1 | - | 1 - 2 | 110 | 67 | ||
Set Membership: |
|