Search
Search

alpha-Man-IIa| Gene Symbol: alpha-Man-IIa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275484.1 | NP_001262413.1 | 2 - 3 | 788 | 93 | |
| NM_079567.3 | NP_524291.2 | 2 - 3 | 785 | 93 | ||
| GenBank mRNA | AY119464.1 | - | 2 - 3 | 718 | 93 | |
Set Membership: |
|