Search
Search

nerfin-2| Gene Symbol: nerfin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275517.1 | NP_001262446.1 | 2 - 3 | 1725 | 64 | |
| NM_079576.3 | NP_524300.1 | 2 - 3 | 1725 | 64 | ||
Set Membership: |
|