Search
Search

Tm1| Gene Symbol: Tm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079636.4 | NP_524360.2 | 7 - 8 | 1110 | 61 | |
| NM_206494.3 | NP_996216.1 | 9 - 10 | 858 | 61 | ||
| GenBank mRNA | AY060414.1 | - | 7 - 8 | 1090 | 61 | |
Set Membership: |
|