Search
Search

osa| Gene Symbol: osa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170168.2 | NP_001163639.1 | 14 - 15 | 4701 | 63 | |
| NM_001275751.1 | NP_001262680.1 | 13 - 14 | 4218 | 63 | ||
| NM_001275752.1 | NP_001262681.1 | 13 - 14 | 4521 | 63 | ||
| NM_079668.3 | NP_524392.2 | 14 - 15 | 4992 | 63 | ||
| NM_169775.2 | NP_732263.1 | 14 - 15 | 4953 | 63 | ||
| NM_206506.2 | NP_996228.1 | 13 - 14 | 4512 | 63 | ||
Set Membership: |
|