Search
Search

vib| Gene Symbol: vib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275797.1 | NP_001262726.1 | 8 - 9 | 892 | 57 | |
| NM_001300473.1 | NP_001287402.1 | 8 - 9 | 796 | 57 | ||
| NM_079680.2 | NP_524404.1 | 8 - 9 | 889 | 57 | ||
| GenBank mRNA | AF246993.1 | - | 8 - 9 | 854 | 57 | |
| BT083455.1 | - | 8 - 9 | 796 | 57 | ||
Set Membership: |
|