Search
Search

Hs6st| Gene Symbol: Hs6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275821.1 | NP_001262750.1 | 4 - 5 | 1664 | 68 | |
| NM_079685.3 | NP_524409.2 | 4 - 5 | 1664 | 68 | ||
| GenBank mRNA | AY051902.1 | - | 4 - 5 | 1641 | 68 | |
Set Membership: |
|