Search
Search

beta-Man| Gene Symbol: beta-Man | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259970.2 | NP_001246899.1 | 8 - 9 | 2185 | 86 | |
| NM_001275312.1 | NP_001262241.1 | 7 - 8 | 2336 | 86 | ||
| NM_001275313.1 | NP_001262242.1 | 7 - 8 | 2298 | 86 | ||
| NM_141179.4 | NP_649436.2 | 7 - 8 | 2298 | 86 | ||
| NM_164315.4 | NP_730783.3 | 6 - 7 | 2370 | 86 | ||
| GenBank mRNA | BT044157.1 | - | 7 - 8 | 2197 | 86 | |
Set Membership: |
|