Search
Search

Prosbeta7| Gene Symbol: Prosbeta7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141272.1 | NP_649529.1 | 1 - 2 | 440 | 69 | |
| NM_169049.2 | NP_730922.1 | 1 - 2 | 251 | 69 | ||
| GenBank mRNA | AY118936.1 | - | 1 - 2 | 208 | 69 | |
Set Membership: |
|