Search
Search

Rpn5| Gene Symbol: Rpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141331.3 | NP_649588.1 | 3 - 4 | 1427 | 67 | |
| GenBank mRNA | AF145315.1 | - | 2 - 3 | 1203 | 67 | |
| AY058339.1 | - | 3 - 4 | 1376 | 67 | ||
Set Membership: |
|