Search
Search

Osi2| Gene Symbol: Osi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275374.1 | NP_001262303.1 | 4 - 5 | 748 | 118 | |
| NM_141364.2 | NP_649621.1 | 4 - 5 | 806 | 118 | ||
| NM_169130.2 | NP_731052.2 | 4 - 5 | 831 | 118 | ||
| GenBank mRNA | BT015251.1 | - | 4 - 5 | 720 | 118 | |
Set Membership: |
|