Search
Search

Osi4| Gene Symbol: Osi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275376.1 | NP_001262305.1 | 2 - 3 | 928 | 85 | |
| NM_141366.2 | NP_649623.2 | 2 - 3 | 928 | 85 | ||
| GenBank mRNA | AY119041.1 | - | 2 - 3 | 928 | 85 | |
Set Membership: |
|