Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02140205_g1
          See other FOXO GE Assays ›
          Gene Symbol:
          foxo
          Gene Name:
          forkhead box, sub-group O
          Gene Aliases:
          3143, Afx, CG3143, DFOXO, DfoxO, Dfoxo, DmQ95V5, Dmel\CG3143, Dmel_CG3143, FKHR, FOXO, FOXO1, FoxO, Foxo, Foxo1, Q95V55, anon-WO0118547.374, cg3143, dFOXO, dFoxO, dFoxo, dfoxO, dfoxo, fd88A, foxO, foxo|dFOXO
          Chromosome Location:
          Chr.3R: 14056945 - 14087978 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0038197
          Assay ID Dm02140205_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: foxo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001275628.1 NP_001262557.1 6 - 7 1456 74
          NM_001316497.1 NP_001303426.1 7 - 8 1599 74
          NM_142073.5 NP_650330.3 7 - 8 1714 74
          NM_206482.3 NP_996204.1 7 - 8 1581 74
          NM_206483.3 NP_996205.1 7 - 8 1972 74
          GenBank mRNA AF426831.1 - 7 - 8 1564 74
          AY089542.1 - 7 - 8 1949 74

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          compound eye morphogenesis
          regulation of transcription, DNA-templated
          response to oxidative stress
          circadian rhythm
          negative regulation of cell proliferation
          insulin receptor signaling pathway
          determination of adult lifespan
          cellular response to starvation
          response to bacterium
          multicellular organism aging
          regulation of autophagy
          positive regulation of autophagy
          negative regulation of lipid storage
          positive regulation of triglyceride catabolic process
          regulation of macroautophagy
          regulation of lipid metabolic process
          negative regulation of cell growth
          cellular response to amino acid starvation
          cellular response to oxidative stress
          regulation of hemocyte proliferation
          female germ-line stem cell population maintenance
          regulation of growth
          regulation of multicellular organism growth
          negative regulation of multicellular organism growth
          glucose homeostasis
          response to starvation
          locomotor rhythm
          negative regulation of cell size
          positive regulation of transcription from RNA polymerase II promoter
          regulation of organ growth
          negative regulation of insulin receptor signaling pathway
          lymph gland development
          dendrite morphogenesis
          negative regulation of wound healing
          positive regulation of entry into reproductive diapause
          negative regulation of fat cell proliferation
          positive regulation of cell cycle arrest
          positive regulation of octopamine signaling pathway
          positive regulation of feeding behavior
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline