Search
Search

foxo| Gene Symbol: foxo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316497.1 | NP_001303426.1 | 8 - 9 | 1783 | 106 | |
| NM_206482.3 | NP_996204.1 | 8 - 9 | 1765 | 106 | ||
| NM_206483.3 | NP_996205.1 | 8 - 9 | 2156 | 106 | ||
| GenBank mRNA | AF426831.1 | - | 8 - 9 | 1748 | 106 | |
| AY089542.1 | - | 8 - 9 | 2133 | 106 | ||
Set Membership: |
|