Search
Search

Mf| Gene Symbol: Mf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170138.1 | NP_001163609.1 | 7 - 8 | 1097 | 112 | |
| NM_001170142.1 | NP_001163613.1 | 7 - 8 | 1031 | 112 | ||
| NM_142166.2 | NP_650423.1 | 7 - 8 | 1187 | 112 | ||
| GenBank mRNA | AY071198.1 | - | 7 - 8 | 1187 | 112 | |
| BT081982.1 | - | 7 - 8 | 1088 | 112 | ||
Set Membership: |
|