Search
Search

Irc| Gene Symbol: Irc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275724.1 | NP_001262653.1 | 7 - 8 | 1809 | 75 | |
| NM_142327.3 | NP_650584.1 | 7 - 8 | 1809 | 75 | ||
| GenBank mRNA | AY051443.1 | - | 7 - 8 | 1787 | 75 | |
Set Membership: |
|