Search
Search

Rim| Gene Symbol: Rim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014630.4 | NP_001014630.3 | 8 - 9 | 3961 | 61 | |
| NM_001260231.1 | NP_001247160.1 | 8 - 9 | 3961 | 61 | ||
| NM_001260232.2 | NP_001247161.2 | 8 - 9 | 3961 | 61 | ||
| NM_001260234.1 | NP_001247163.1 | 8 - 9 | 3961 | 61 | ||
| NM_001260237.2 | NP_001247166.2 | 8 - 9 | 3961 | 61 | ||
| NM_001260238.2 | NP_001247167.2 | 8 - 9 | 3961 | 61 | ||
| NM_001260239.1 | NP_001247168.1 | 8 - 9 | 3961 | 61 | ||
| NM_001260240.2 | NP_001247169.2 | 8 - 9 | 3961 | 61 | ||
| NM_001260241.2 | NP_001247170.2 | 8 - 9 | 3961 | 61 | ||
Set Membership: |
|