Search
Search

wrd| Gene Symbol: wrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144598.3 | NP_001138070.1 | 4 - 5 | 927 | 64 | |
| NM_001144599.2 | NP_001138071.1 | 4 - 5 | 927 | 64 | ||
| NM_001144600.2 | NP_001138072.1 | 2 - 3 | 310 | 64 | ||
| NM_001260247.1 | NP_001247176.1 | 4 - 5 | 1059 | 64 | ||
| NM_001260248.1 | NP_001247177.1 | 4 - 5 | 886 | 64 | ||
| NM_001260249.1 | NP_001247178.1 | 4 - 5 | 1333 | 64 | ||
| NM_001260250.1 | NP_001247179.1 | 3 - 4 | 2032 | 64 | ||
| NM_001275765.1 | NP_001262694.1 | 5 - 6 | 1476 | 64 | ||
| NM_001275766.1 | NP_001262695.1 | 5 - 6 | 1476 | 64 | ||
| NM_142424.3 | NP_650681.2 | 4 - 5 | 1100 | 64 | ||
| NM_169787.2 | NP_732293.1 | 4 - 5 | 927 | 64 | ||
| NM_169788.1 | NP_732294.1 | 2 - 3 | 429 | 64 | ||
| NM_169789.2 | NP_732295.1 | 4 - 5 | 1875 | 64 | ||
| NM_169790.1 | NP_732296.1 | 4 - 5 | 1875 | 64 | ||
| GenBank mRNA | AY118546.1 | - | 4 - 5 | 1875 | 64 | |
| BT021232.1 | - | 4 - 5 | 926 | 64 | ||
Set Membership: |
|