Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02141794_g1
          See other WRD GE Assays ›
          Gene Symbol:
          wrd
          Gene Name:
          well-rounded
          Gene Aliases:
          B', B'/PR61, B56-1, BcDNA:GM05554, CG 7913, CG7901, CG7913, Dmel\CG7913, Dmel_CG7913, PP2A, PP2A B', PP2A-B, PP2A-B', PP2A[B'-1], Wrd, anon-WO0118547.420, dB56-1, dPP2A, dPP2A-B56-1, i234, pp2A-B'
          Chromosome Location:
          Chr.3R: 18167757 - 18180049 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0042693
          Assay ID Dm02141794_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: wrd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144598.3 NP_001138070.1 7 - 8 1884 60
          NM_001144599.2 NP_001138071.1 7 - 8 1884 60
          NM_001144600.2 NP_001138072.1 5 - 6 1267 60
          NM_001260247.1 NP_001247176.1 7 - 8 2016 60
          NM_001260248.1 NP_001247177.1 7 - 8 1843 60
          NM_001260249.1 NP_001247178.1 7 - 8 2290 60
          NM_001260250.1 NP_001247179.1 6 - 7 2989 60
          NM_001275765.1 NP_001262694.1 8 - 9 2433 60
          NM_001275766.1 NP_001262695.1 8 - 9 2433 60
          NM_142424.3 NP_650681.2 7 - 8 2057 60
          NM_169787.2 NP_732293.1 7 - 8 1884 60
          NM_169788.1 NP_732294.1 5 - 6 1386 60
          NM_169789.2 NP_732295.1 7 - 8 2832 60
          NM_169790.1 NP_732296.1 7 - 8 2832 60
          GenBank mRNA AY118546.1 - 7 - 8 2832 60
          BT021232.1 - 7 - 8 1883 60

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          autophagy
          neuromuscular synaptic transmission
          neuromuscular junction development
          regulation of protein autophosphorylation
          negative regulation of apoptotic process
          sex comb development
          negative regulation of insulin receptor signaling pathway
          triglyceride homeostasis
          synaptic vesicle localization
          negative regulation of TORC1 signaling
          protein phosphatase regulator activity
          protein phosphatase activator activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline