Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02142721_g1
          See other SYP GE Assays ›
          Gene Symbol:
          Syp
          Gene Name:
          Syncrip
          Gene Aliases:
          AI945337, CG17838, Dmel\CG17838, Dmel_CG17838, anon-WO0118547.613, cg17838, l(3)03806, syp
          Chromosome Location:
          Chr.3R: 20756729 - 20810726 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0038826
          Assay ID Dm02142721_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104378.2 NP_001097848.2 3 - 4 1033 81
          NM_001104379.2 NP_001097849.1 3 - 4 956 81
          NM_001275847.1 NP_001262776.1 4 - 5 1043 81
          NM_001275848.1 NP_001262777.1 3 - 4 782 81
          NM_001275849.1 NP_001262778.1 4 - 5 1043 81
          NM_001275850.1 NP_001262779.1 4 - 5 1043 81
          NM_001275851.1 NP_001262780.1 3 - 4 2368 81
          NM_001275852.1 NP_001262781.1 3 - 4 782 81
          NM_001275853.1 NP_001262782.1 3 - 4 1033 81
          NM_001275854.1 NP_001262783.1 3 - 4 782 81
          NM_001275855.1 NP_001262784.1 3 - 4 2368 81
          NM_001369986.1 NP_001356920.1 4 - 5 1043 81
          NM_001369987.1 NP_001356921.1 4 - 5 1043 81
          NM_001369988.1 NP_001356922.1 3 - 4 782 81
          NM_142656.4 NP_650913.2 3 - 4 2368 81
          NM_169925.3 NP_732555.1 3 - 4 782 81
          NM_169926.2 NP_732556.1 3 - 4 782 81
          NM_169927.3 NP_732557.2 4 - 5 1043 81
          NM_169928.3 NP_732558.2 3 - 4 1482 81
          NM_169929.3 NP_732559.1 3 - 4 1033 81
          GenBank mRNA AY058477.1 - 3 - 4 1331 81

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          oocyte dorsal/ventral axis specification
          negative regulation of synaptic growth at neuromuscular junction
          regulation of oskar mRNA translation
          dorsal appendage formation
          regulation of neuromuscular synaptic transmission
          RNA binding
          mRNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline