Search
Search

Nmnat| Gene Symbol: Nmnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276013.2 | NP_001262942.1 | 5 - 6 | 1111 | 65 | |
| NM_143058.1 | NP_651315.2 | 5 - 6 | 1147 | 65 | ||
| GenBank mRNA | BT030750.1 | - | 5 - 6 | 1146 | 65 | |
Set Membership: |
|