Search
Search

Dak1| Gene Symbol: Dak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143199.3 | NP_651456.1 | 1 | 987 | 149 | |
| GenBank mRNA | AY119622.1 | - | 1 | 955 | 149 | |
| AY296751.1 | - | 1 | 950 | 149 | ||
Set Membership: |
|