Search
Search

Tusp| Gene Symbol: Tusp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202374.2 | NP_001189303.1 | 10 - 11 | 4974 | 82 | |
| NM_001276085.2 | NP_001263014.1 | 10 - 11 | 4611 | 82 | ||
| NM_001276086.2 | NP_001263015.1 | 10 - 11 | 4602 | 82 | ||
| NM_143316.4 | NP_651573.4 | 10 - 11 | 4374 | 82 | ||
| GenBank mRNA | BT132902.1 | - | 10 - 11 | 4374 | 82 | |
Set Membership: |
|