Search
Search

tau| Gene Symbol: tau | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276088.1 | NP_001263017.1 | 5 - 6 | 1414 | 90 | |
| NM_001276089.1 | NP_001263018.1 | 5 - 6 | 1232 | 90 | ||
| NM_001300634.1 | NP_001287563.1 | 5 - 6 | 1232 | 90 | ||
| NM_001300637.1 | NP_001287566.1 | 5 - 6 | 770 | 90 | ||
| NM_001300638.1 | NP_001287567.1 | 4 - 5 | 792 | 90 | ||
| NM_001316620.1 | NP_001303549.1 | 5 - 6 | 1232 | 90 | ||
| NM_143318.3 | NP_651575.1 | 5 - 6 | 1232 | 90 | ||
| NM_170345.3 | NP_733224.2 | 7 - 8 | 2743 | 90 | ||
| GenBank mRNA | AY032977.1 | - | 5 - 6 | 1209 | 90 | |
| AY102697.1 | - | 5 - 6 | 1228 | 90 | ||
Set Membership: |
|