Search
Search

alpha-Man-Ib| Gene Symbol: alpha-Man-Ib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143410.4 | NP_651667.1 | 2 - 3 | 676 | 82 | |
| GenBank mRNA | AY058775.1 | - | 2 - 3 | 658 | 82 | |
Set Membership: |
|