Search
Search

CG31445| Gene Symbol: CG31445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276135.1 | NP_001263064.1 | 3 - 4 | 821 | 64 | |
| NM_143432.3 | NP_651689.1 | 3 - 4 | 683 | 64 | ||
| GenBank mRNA | AY060643.1 | - | 3 - 4 | 663 | 64 | |
Set Membership: |
|