Search
Search

CG1815| Gene Symbol: CG1815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143624.3 | NP_651881.2 | 4 - 5 | 1540 | 67 | |
| NM_170563.2 | NP_733442.1 | 4 - 5 | 1528 | 67 | ||
| GenBank mRNA | BT010115.1 | - | 4 - 5 | 1540 | 67 | |
Set Membership: |
|