Search
Search

mtd| Gene Symbol: mtd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031977.3 | NP_001027148.1 | 4 - 5 | 1175 | 69 | |
| NM_001043213.3 | NP_001036678.1 | 4 - 5 | 1202 | 69 | ||
| NM_001259999.2 | NP_001246928.1 | 4 - 5 | 1428 | 69 | ||
| NM_143760.4 | NP_652017.1 | 4 - 5 | 1202 | 69 | ||
| NM_169038.3 | NP_730909.1 | 4 - 5 | 1128 | 69 | ||
| GenBank mRNA | AF125384.1 | - | 4 - 5 | 1200 | 69 | |
Set Membership: |
|