Search
Search

sqd| Gene Symbol: sqd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260159.2 | NP_001247088.1 | 4 - 5 | 1116 | 104 | |
| NM_143952.2 | NP_652209.1 | 4 - 5 | 841 | 104 | ||
| NM_169528.2 | NP_731825.1 | 4 - 5 | 1116 | 104 | ||
| NM_169529.3 | NP_731826.1 | 4 - 5 | 845 | 104 | ||
| NM_206481.2 | NP_996203.1 | 5 - 6 | 1328 | 104 | ||
| GenBank mRNA | AY118501.1 | - | 4 - 5 | 1096 | 104 | |
| BT001384.1 | - | 5 - 6 | 1048 | 104 | ||
| BT003283.1 | - | 5 - 6 | 1301 | 104 | ||
| BT023832.1 | - | 4 - 5 | 824 | 104 | ||
| X62637.1 | - | 825 | 104 | |||
Set Membership: |
|