Search
Search

SF2| Gene Symbol: SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260210.2 | NP_001247139.1 | 2 - 3 | 635 | 108 | |
| NM_144354.3 | NP_652611.1 | 2 - 3 | 635 | 108 | ||
| GenBank mRNA | AF232773.1 | - | 2 - 3 | 619 | 108 | |
| AY051919.1 | - | 2 - 3 | 619 | 108 | ||
Set Membership: |
|