Search
Search

mod(mdg4)
pre-mod(mdg4)-B
pre-mod(mdg4)-G
pre-mod(mdg4)-H
pre-mod(mdg4)-I
pre-mod(mdg4)-J
pre-mod(mdg4)-K
pre-mod(mdg4)-L
pre-mod(mdg4)-P| Gene Symbol: mod(mdg4) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_163886.2 | NP_732628.1 | 5 - 6 | 1564 | 56 | |
| GenBank mRNA | BT003579.1 | - | 2467 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300520.2 | NP_001287449.1 | 2 - 3 | 1470 | 56 | |
| NM_001316584.1 | NP_001303513.1 | 2 - 3 | 1470 | 56 | ||
| NM_001316585.1 | NP_001303514.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316586.1 | NP_001303515.1 | 1 - 2 | 1763 | 56 | |
| NM_001316587.1 | NP_001303516.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316582.1 | NP_001303511.1 | 2 - 3 | 1568 | 56 | |
| NM_001316583.1 | NP_001303512.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300517.1 | NP_001287446.1 | 2 - 3 | 1248 | 56 | |
| NM_001316592.1 | NP_001303521.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300519.1 | NP_001287448.1 | 2 - 3 | 775 | 56 | |
| NM_001316588.1 | NP_001303517.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300518.1 | NP_001287447.1 | 1 - 2 | 1763 | 56 | |
| NM_001316590.1 | NP_001303519.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300516.1 | NP_001287445.1 | 2 - 3 | 1607 | 56 | |
| NM_001316591.1 | NP_001303520.1 | 1 - 2 | 1763 | 56 | ||
| Gene Symbol: pre-mod(mdg4)-P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300515.1 | NP_001287444.1 | 2 - 3 | 1470 | 56 | |
| NM_001316589.1 | NP_001303518.1 | 1 - 2 | 1763 | 56 | ||
Set Membership: |
|