Search
Search

Alh| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104227.3 | NP_001097697.2 | 6 - 7 | 3247 | 137 | |
| NM_001170065.2 | NP_001163536.1 | 6 - 7 | 3247 | 137 | ||
| NM_001170066.2 | NP_001163537.1 | 6 - 7 | 3247 | 137 | ||
| NM_169170.3 | NP_731131.2 | 6 - 7 | 3247 | 137 | ||
Set Membership: |
|