Search
Search

Alh| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104227.3 | NP_001097697.2 | 9 - 10 | 4503 | 66 | |
| NM_001170065.2 | NP_001163536.1 | 9 - 10 | 4503 | 66 | ||
| NM_001170066.2 | NP_001163537.1 | 9 - 10 | 4410 | 66 | ||
| NM_001275401.1 | NP_001262330.1 | 9 - 10 | 4183 | 66 | ||
| NM_001275402.1 | NP_001262331.1 | 9 - 10 | 4407 | 66 | ||
| NM_169170.3 | NP_731131.2 | 9 - 10 | 4279 | 66 | ||
Set Membership: |
|