Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PpD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275504.2 | NP_001262433.1 | 4 - 5 | 795 | 76 | |
NM_080207.3 | NP_524946.1 | 5 - 6 | 900 | 76 | ||
NM_169307.2 | NP_731398.1 | 5 - 6 | 765 | 76 | ||
GenBank mRNA | AJ271781.1 | - | 4 - 5 | 714 | 76 | |
AY058356.1 | - | 4 - 5 | 758 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |