Search
Search

Ubx| Gene Symbol: Ubx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080504.4 | NP_536752.1 | 2 - 3 | 1786 | 61 | |
| NM_169729.3 | NP_732172.1 | 2 - 3 | 1759 | 61 | ||
| GenBank mRNA | BT010241.1 | - | 2 - 3 | 1759 | 61 | |
| X76210.1 | - | 1986 | 61 | |||
Set Membership: |
|