Search
Search

Stat92E| Gene Symbol: Stat92E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014643.2 | NP_001014643.1 | 6 - 7 | 2497 | 66 | |
| NM_001275828.1 | NP_001262757.1 | 6 - 7 | 2341 | 66 | ||
| NM_001275833.1 | NP_001262762.1 | 7 - 8 | 2910 | 66 | ||
| NM_169899.3 | NP_732516.2 | 7 - 8 | 2889 | 66 | ||
| NM_169900.3 | NP_732517.2 | 6 - 7 | 2476 | 66 | ||
| NM_206520.2 | NP_996242.1 | 7 - 8 | 2733 | 66 | ||
| NM_206521.2 | NP_996243.1 | 7 - 8 | 2754 | 66 | ||
| GenBank mRNA | BT021314.1 | - | 7 - 8 | 2755 | 66 | |
| U40070.1 | - | 7 - 8 | 2729 | 66 | ||
Set Membership: |
|