Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02150103_m1
          See other ATPALPHA GE Assays ›
          Gene Symbol:
          Atpalpha
          Gene Name:
          Na pump alpha subunit
          Gene Aliases:
          (Na[+]/k[+])-ATPase, ATP-alpha, ATPBagr, ATPa, ATPalpha, Atp, Atp a, Atpa, CG5670, Da-47, Dmel\CG5670, Dmel_CG5670, JY, K ATPase, K-ATPase, K[+] -ATPase, K[+]-ATPase, K[+]-ATPase alpha subunit, K[+]-ATPase alpha-subunit, NKA, Na, Na+-K+-ATPase, Na+/K+ ATPase alpha subunit, Na+/K+-ATPase, Na-p, Na/K ATPase, Na/K-ATPase, Na/K-ATPase alpha-subunit, NaK, NaK ATPase, NaK-ATPase, Na[+], Na[+]-K[+]-ATPase alpha[[5]]-subunit, Na[+]/K[+] ATPase, Na[+]/K[+]-ATPase, Na[+]/K[+]ATPase, Na[+]K[+] ATPase, Na[+]K[+]-ATPase, Na[+]K[+]ATPase, alpha-ATPase, atpalpha, dnATPase, l(3)01164, l(3)01453, l(3)04492, l(3)j3B2, l(3)j3B7, l(3)j6A4, l(3)rM454, l(3)ry136, sodium/potassium transporting ATPase alpha
          Chromosome Location:
          Chr.3R: 20948714 - 20976239 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0002921
          Assay ID Dm02150103_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atpalpha
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170196.3 NP_001163667.1 7 - 8 2642 79
          NM_169939.2 NP_732575.1 6 - 7 3217 79
          GenBank mRNA BT044212.1 - 6 - 7 3190 79

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          cation transport
          cellular sodium ion homeostasis
          chemical synaptic transmission
          sensory perception of sound
          locomotory behavior
          jump response
          determination of adult lifespan
          adult locomotory behavior
          response to temperature stimulus
          response to mechanical stimulus
          septate junction assembly
          cellular potassium ion homeostasis
          regulation of tube diameter, open tracheal system
          regulation of tube length, open tracheal system
          sodium ion export from cell
          neuromuscular process
          synaptic growth at neuromuscular junction
          trachea morphogenesis
          hydrogen ion transmembrane transport
          potassium ion import across plasma membrane
          sodium:potassium-exchanging ATPase activity
          ATP binding
          cation transmembrane transporter activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline