Search
Search

Rox8| Gene Symbol: Rox8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275968.1 | NP_001262897.1 | 4 - 5 | 2241 | 61 | |
| NM_001275969.1 | NP_001262898.1 | 4 - 5 | 2283 | 61 | ||
| NM_170118.2 | NP_732944.2 | 4 - 5 | 2135 | 61 | ||
| NM_170119.2 | NP_732945.1 | 4 - 5 | 2213 | 61 | ||
| GenBank mRNA | BT050464.1 | - | 4 - 5 | 2213 | 61 | |
Set Membership: |
|