Search
Search

Nf1| Gene Symbol: Nf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014668.2 | NP_001014668.1 | 1 - 2 | 337 | 55 | |
| NM_001276049.1 | NP_001262978.1 | 1 - 2 | 337 | 55 | ||
| NM_001276050.1 | NP_001262979.1 | 1 - 2 | 337 | 55 | ||
| NM_170252.3 | NP_733131.2 | 1 - 2 | 337 | 55 | ||
| NM_170253.2 | NP_733132.2 | 1 - 2 | 337 | 55 | ||
Set Membership: |
|