Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02151072_g1
          See other GRO GE Assays ›
          Gene Symbol:
          gro
          Gene Name:
          groucho
          Gene Aliases:
          BEST:GM01575, BEST:LD15161, BcDNA.LD33829, BcDNA:LD33829, CG8384, DGro, Dmel\CG8384, Dmel_CG8384, E(spl)-WD, E(spl)[2], E(spl)gro, E(spl)m9/m10, En-spl, GRO, Grg/TLE, Gro, Groucho, anon-WO0118547.385, dAES1, dAES2, dGro, l(3)gro, m10, m9, m9-m10, m9/10, m9/m10
          Chromosome Location:
          Chr.3R: 26041736 - 26049911 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0001139
          Assay ID Dm02151072_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: gro
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001202364.1 NP_001189293.1 12 - 13 2576 58
          NM_001202365.2 NP_001189294.1 13 - 14 2597 58
          NM_001260380.2 NP_001247309.1 12 - 13 2540 58
          NM_001276052.1 NP_001262981.1 11 - 12 2507 58
          NM_001300610.1 NP_001287539.1 12 - 13 2564 58
          NM_079790.5 NP_524514.2 12 - 13 2509 58
          NM_170254.3 NP_733133.1 12 - 13 2589 58
          NM_170255.4 NP_733134.1 12 - 13 2570 58
          NM_170256.2 NP_733135.1 12 - 13 2488 58
          NM_206575.3 NP_996298.1 13 - 14 2603 58
          GenBank mRNA AF145695.1 - 12 - 13 2555 58

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription, DNA-templated
          epidermal growth factor receptor signaling pathway
          nervous system development
          sex determination, primary response to X:A ratio
          torso signaling pathway
          fibroblast growth factor receptor signaling pathway
          negative regulation of Toll signaling pathway
          negative regulation of transcription, DNA-templated
          negative regulation of canonical Wnt signaling pathway
          transcription corepressor activity
          protein binding
          transcription factor binding
          repressing transcription factor binding
          HMG box domain binding
          CRD domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline