Search
Search

Pli| Gene Symbol: Pli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104420.3 | NP_001097890.1 | 5 - 6 | 934 | 63 | |
| NM_079742.4 | NP_524466.1 | 5 - 6 | 1006 | 63 | ||
| GenBank mRNA | AF091624.1 | - | 5 - 6 | 911 | 63 | |
| AY069632.1 | - | 5 - 6 | 982 | 63 | ||
Set Membership: |
|