Search
Search

Rab7| Gene Symbol: Rab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260347.1 | NP_001247276.1 | 3 - 4 | 676 | 99 | |
| NM_001260348.2 | NP_001247277.1 | 3 - 4 | 687 | 99 | ||
| NM_079748.4 | NP_524472.1 | 3 - 4 | 722 | 99 | ||
| GenBank mRNA | AB035672.1 | - | 3 - 4 | 708 | 99 | |
| AF263363.1 | - | 3 - 4 | 702 | 99 | ||
| AY060236.1 | - | 3 - 4 | 702 | 99 | ||
| BT124755.1 | - | 3 - 4 | 676 | 99 | ||
Set Membership: |
|