Search
Search

nAChRalpha1| Gene Symbol: nAChRalpha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275987.1 | NP_001262916.1 | 6 - 7 | 2002 | 61 | |
| NM_001275988.2 | NP_001262917.1 | 6 - 7 | 2002 | 61 | ||
| NM_079757.4 | NP_524481.2 | 6 - 7 | 2002 | 61 | ||
Set Membership: |
|