Search
Search

fkh| Gene Symbol: fkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170291.1 | NP_001163762.1 | 1 | 3152 | 141 | |
| NM_001276109.1 | NP_001263038.1 | 1 | 3152 | 141 | ||
| NM_001300645.1 | NP_001287574.1 | 1 | 3304 | 141 | ||
| NM_001316532.1 | NP_001303461.1 | 1 | 3152 | 141 | ||
| NM_079818.3 | NP_524542.1 | 1 | 3152 | 141 | ||
| GenBank mRNA | BT150445.1 | - | 1 | 3104 | 141 | |
Set Membership: |
|